Learn More
STEMCELL Technologies Human Recombinant Oncostatin M, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78094.2
Encompass_Preferred
Use oncostatin M (OSM) to modulate differentiation, cell proliferation, hematopoiesis, and inflammation (Tanaka & Miyajima. Rev Physiol Biochem Pharmacol, 2003). OSM, a member of interleukin 6 family of cytokines, bears close resemblance to both leukemia-inhibitory factor and granulocyte colony-stimulating factor in amino acid sequence and their modulation of differentiation in a variety of cell types (Rose & Bruce. Proc Natl Acad Sci USA, 1991). OSM activates the JAK/STAT pathway by signaling through type I receptor (consisting of gp130 and LIF receptor) and type II receptor (consisting of gp130 and OSM receptor) (Auguste et al. J Biol Chem, 1997; Gómez-Lechón. Life Sci, 1999). OSM is primarily produced by activated T cells and monocytes, as well as activated macrophages, neutrophils, mast cells, and dendritic cells. Human recombinant Oncostatin M from STEMCELL comes lyophilized with ≥ 95% purity; EC50 ≤ 10 ng/mL; endotoxin levels verified ≤ 0.2 EU/μg protein. Alternative names: OSM.
Catalog No. 50-226-9656
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.