Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OncoStat Panel, SNAP-ChIP™ Spike-in
SDP

SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.

Supplier:  Invitrogen™ A47355

 View more versions of this product

Catalog No. A47355


Explore all special offers
Add to Cart

Description

Description

  • SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
  • Homogenous, fully defined standards faithfully represent target mononucleosomes in the experimental sample
  • Nucleosomes are subjected to rigorous quality control for lot-to-lot consistency
  • Unique DNA barcodes can be distinguished from experimental sample genomes
  • Spike-ins provide a direct readout of antibody performance in ChIP
  • Ability to monitor technical variability between samples
  • Defined standards enable universal normalization across experiments
  • SNAP-ChIP panels are directly compatible with your current ChIP workflow, with semi-synthetic nucleosomes bearing the mutation of interest immunoprecipitated and processed alongside sample chromatin.
  • Users can directly monitor antibody capability (specificity and enrichment) in a ChIP experiment by measuring the recovery of on- and off-target nucleosomes. The SNAP spike-ins can be further used as a defined standard to normalize your ChIP data, thereby controlling for technical variability across experiments.
  • The OncoStat Panel consists of a pool of nucleosomes harboring seven well-studied histone H3.3 mutations that have been implicated in cancer (histone H3.3K4M, H3.3K9M, H3.3K27M, H3.3G34R, H3.3G34V, H3.3G34W and H3.3K36M) plus a wild-type H3.3 control. Further, each of these eight nucleosomes is wrapped by two independent DNA barcodes (16 total barcodes in the panel), allowing for an internal technical replicate in each ChIP reaction.
  • A single spike-in of the panel allows users to check antibody specificity by examining the post-IP recovery of on- versus off-target SNAP-ChIP nucleosomes, supporting the generation of high quality ChIP data.
Specifications

Specifications

SNAP-ChIP Panel
Recombinant Nucleosome Panel
OncoStat™, SNAP-ChIP™
Store at -5 to -30°C.
100 Reactions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.