Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OncoStat Panel, SNAP-ChIP™ Spike-in
SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
Supplier: Invitrogen™ A47343
Description
- SNAP-ChIP (Sample Normalization and Antibody Profiling Chromatin ImmunoPrecipitation) uses DNA-barcoded semi-synthetic nucleosomes bearing distinct histone mutations as spike-in ChIP controls.
- Homogenous, fully defined standards faithfully represent target mononucleosomes in the experimental sample
- Nucleosomes are subjected to rigorous quality control for lot-to-lot consistency
- Unique DNA barcodes can be distinguished from experimental sample genomes
- Spike-ins provide a direct readout of antibody performance in ChIP
- Ability to monitor technical variability between samples
- Defined standards enable universal normalization across experiments
- SNAP-ChIP panels are directly compatible with your current ChIP workflow, with semi-synthetic nucleosomes bearing the mutation of interest immunoprecipitated and processed alongside sample chromatin.
- Users can directly monitor antibody capability (specificity and enrichment) in a ChIP experiment by measuring the recovery of on- and off-target nucleosomes. The SNAP spike-ins can be further used as a defined standard to normalize your ChIP data, thereby controlling for technical variability across experiments.
- The OncoStat Panel consists of a pool of nucleosomes harboring seven well-studied histone H3.3 mutations that have been implicated in cancer (histone H3.3K4M, H3.3K9M, H3.3K27M, H3.3G34R, H3.3G34V, H3.3G34W and H3.3K36M) plus a wild-type H3.3 control. Further, each of these eight nucleosomes is wrapped by two independent DNA barcodes (16 total barcodes in the panel), allowing for an internal technical replicate in each ChIP reaction.
- A single spike-in of the panel allows users to check antibody specificity by examining the post-IP recovery of on- versus off-target SNAP-ChIP nucleosomes, supporting the generation of high quality ChIP data.
Specifications
Store at -5 to -30°C. | |
10 Reactions | |
SNAP-ChIP Panel |
Recombinant Nucleosome Panel | |
OncoStat™, SNAP-ChIP™ |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction