Learn More
Cytiva (Formerly Pall Lab) Omega™ Membrane Discs - 30K - 62 mm (12/pkg)
Supplier: Cytiva OM030062
High Performance and Stability for Users of Conventional, Non-Disposable Stirred Cell-Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose. This results in lower processing times and the highest possible recoveries.-Discs can be washed and reused. This product is not approved for use in medical, clinical, surgical or other patient applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.