Learn More
Enzo Life Sciences ODN 2395 (TLRGRADE) (synthetic) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX746020C100
Encompass_Preferred
TLR9 ligand. CpG oligodeoxynucleotide (Type C) with phosphorothioate backbone. Specific ligand for human and mouse TLR9 (Toll-like receptor 9). MW: 7068 (ammonium salt). Source: Synthetic. Purification: Activity and endotoxin tested - TLRGRADE. Formulation: Lyophilized. Sterile.
Catalog No. 50-210-0192
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.