Learn More
Enzo Life Sciences ODN 1826 (TLRGRADE™) (synthetic) (BULK) (1mg)
Supplier: Enzo Life Sciences ALX746052M001
CpG oligodeoxynucleotide (Type B) with phosphorothioate backbone. Specific ligand for mouse TLR9 (Toll-like receptor 9). Alternative name: CpG-B. Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.