Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
o-Xylene, Puriss. p.a.,≥99.0% (GC), Riedel-de Haen, Honeywell
Click to view available options
Quantity:
1 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
CAS Max % | 100.00 |
Beilstein | 1815558 |
Grade | PA |
Density | 0.88 g/cm3 |
Boiling Point | 143°C to 145°C |
Quantity | 1 L |
Flash Point | 30°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | H226+H304+H312+H332+H315+H319+H335+H336 |
Precautionary Statement | P210+P233+P240+P241+P242+P243+P261+P264+P271+P280 |
Recommended Storage | Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction