Learn More
Selleck Chemical LLC Nystatin-50mg
Supplier: Selleck Chemical LLC S1934
Nystatin(Fungicidin), which belongs to the polyene group of antimycotics, is frequently used as a topical agent in the treatment of oro-pharyngeal candidosis.