Learn More
STEMCELL Technologies Nuclease-Free Water, Size: 2 mL
Supplier: STEMCELL Technologies 79002
Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.