Learn More
Advanced ImmunoChemical, Inc. Human Recombinant NT-PROBNP (Amino Terminal Human Brain Natriuretic Peptide Precursor), 0.05mg.
Supplier: Advanced ImmunoChemical, Inc. 8NTPBRH0.05MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Human recombinant NT-proBNP (amino terminal human brain natriuretic peptide precursor). 0.05mg. Source:Recombinant E.coli.Applications:Calibrator or standard in immunoassays, immunogen for antibody production.Analysis:Purity > 95 % (Tricine-SDS-PAGE).Concentration is determined spectrophotometrically.Immunoreactivity is confirmed by reaction with monoclonal antibodies specific to different fragments of NT-proBNP molecule.Predicted molecular mass 8589 Da confirmed by MALDI-MS.Recombinant human NT-proBNP contains additional methionine residue at the N-terminus (in comparison with native NT-proBNP).Purification:Immunoaffinity purification followed by ion-exchange chromatography.Presentation:10 mM potassium phosphate, pH 7.4, 150 mM NaClStorage Conditions:-70 °C (between -65 and -80 °C allowed)Other information:Avoid repeated freezing and thawing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.