Learn More
Andwin Scientific NS BULK CONTAINERS 4.5OZ 500CS
SDP
Supplier:  Andwin Scientific 25384148
Encompass_Preferred
The Polypropylene Specimen Containers feature durability and graduations in 10 mL and 0.5 oz. increments. This product is constructed with translucent polypropylene to permit users to easily and conveniently view the contents. The container is nonste
Catalog No. NC0789327
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.