Learn More
US Biomax Inc NONSMALL CELL LUNG CANCR(NSCLC
SDP
Supplier:  US Biomax Inc LC819A
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Non-small cell lung cancer (NSCLC) carcinoma tissue array, including pathology grade, TNM / Stage, 80 cases/80 cores(core size 1.5mm),replacing LC819, unstained slide
Catalog No. NC2145892