Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Polyclonal Antibody, HRP, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A150128P
Description
Nitrotyrosine Polyclonal antibody specifically detects Nitrotyrosine in Human samples. It is validated for ELISA, Western BlotSpecifications
Nitrotyrosine | |
Polyclonal | |
HRP | |
Nitro-tyrosine | |
Antigen affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.1M NaCl and 0.01% thimerosal; pH 7.4 | |
Goat | |
500 μg | |
Primary | |
4° C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction