Learn More
Enzo Life Sciences Nitric Oxide (total), detection kit (2x96wells)
Supplier: Enzo Life Sciences ADI917020
The Nitric Oxide (total), detection kit is based on the enzymatic conversion of nitrate to nitrite by the enzyme nitrate reductase, followed by the Griess reaction to form a colored azo dye product. Quantification is performed by measuring absorption at 540-570 nm. Only 0.050 ml diluted sample needed per well saves precious sample. Sensitivity: 0.625 µmol/L (range 3.125 - 100 µmol/L). Applications: Colorimetric detection. Species Reactivity: Species independent. Contents: Microtiter plate, Nitrate Reductase, Nitrate Reductase Storage Buffer, NADH, Nitrate Standard, Reaction buffer concentrate, Griess Reagent I, Griess Reagent II
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.