Learn More
Bioassay Systems QuantiChrom™ Nitric Oxide Assay Kit. For quantitative determination of nitric oxide (nitrate/nitrite) and evaluation of drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems D2NO100
Encompass_Preferred
For quantitative determination of nitric oxide (nitrate/nitrite) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Detection range 0.6 - 200 µM in 96-well plate. Rapid and reliable. Using an optimized VCl3 reagent, the time required for reduction of nitrate (NO3-) to nitrite (NO2-) is reduced to 10 min at 60°C. Simple and high-throughput. The procedure involves mixing sample with three reagents, incubation for 10 min at 60°C and reading the optical density. Can be readily automated to measure thousands of samples per day. Method: OD540nm. Samples: Plasma, serum, urine, tissue/cells and foods. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.6 µM.
Catalog No. 50-489-194
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.