Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific™

Catalog No. p-7138957
Click to view available options
Quantity:
500 mL
2.5 L

Suitable for digestion of samples prior to trace metal analysis via ICP-OES. Tested for 34 metals at ppb levels and also meets ACS testing specifications

  • Suitable for trace metal analysis by ICP-OES
  • Tested for 34 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
  • Offered in 500mL and 2.5L poly coated glass bottles

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance Clear, colorless liquid
Color 10 max. (APHA)
Density 1.42
Heavy Metals (as Pb) 100 ppb max.
Packaging Poly coated glass bottle
Residue after Ignition ≤ 2 ppm
Quantity 500 mL
MDL Number MFCD00011349
Percent Purity 69.0 to 70.0%
Grade For trace metal analysis
Chemical Name or Material Nitric Acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.