Learn More
Glass Expansion Nickel Hyper Skimmer Cone PerkinElmer NexION 5000
Supplier:  Glass Expansion PE4015NI
Encompass_Preferred
Nickel Hyper Skimmer Cone (N8160120) – Standard nickel hyper skimmer cone for PerkinElmer NexION 5000 ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2950
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.