Learn More
Glass Expansion Nickel Skimmer Cone (Robust Plasma) compatible with Thermo Q, RQ and TQ. New insert-free design eliminates need for 4.5mm insert.
Supplier: Glass Expansion TG1045NI
Nickel Skimmer Cone – designed for robust plasma tuning conditions, for Thermo Q/RQ/TQ. Replaces combination of 1311870 and BRE0006591. Design eliminates the need for removable insert. Ideal for long-term analysis of high matrix samples that would normally require a dilution, and helps minimize drift and cone maintenance. Not recommended for samples containing hydrofluoric acid (HF). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1028) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number TG1048-Pt.For high matrix tolerance and high sensitivity, we recommend NC2046959.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.