Learn More
New England Biolabs, Inc. NEBNext Ultra™ DNA Library Prep Kit for Illumina – 96 reactions
Supplier: New England Biolabs, Inc. E7370L
This original version of the Ultra DNA Library Prep Kit enables library preparation from 5 ng – 1 µg of input DNA in a streamlined workflow. However, we now recommend use of the more recent NEBNext Ultra II DNA Library Prep Kit for Illumina, which has been further optimized to provide increased yields of high quality libraries from a broader range of input amounts (500 pg – 1 µg).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.