Learn More
R & D SYSTEMS INC Newborn Calf Serum, Heat Inactivated
SDP
Supplier:  R & D SYSTEMS INC S11210H
Encompass_Preferred
Call your Customer Service Representative for bulk discounts. R&D Systems Newborn Calf Serum (NBC, NBCS) provides many of the same nutrients as Fetal Bovine Serum, but has slightly higher protein levels. For routine maintenance of select cell lines and where low antibody concentrations are not critical, Newborn Calf Serum isoften used as a lower-priced alternative to FBS. Heat Inactivated FBS is treated by heat inactivation where serum is exposed to a temperature of 56 °C for 30 minutes under controlled conditions. All R&D Systems Sera are manufactured in an ISO 9001:2015 certified facility.
Catalog No. 50-152-7071
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.