Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutral Red, 1% AQ, Vion Biosciences
Shop All Aldon Products
Click to view available options
Quantity:
500 mL
Description
- Clearly differentiate between cell and tissue types in histological samples with 1% aqueous neutral red.
Specifications
Specifications
Melting Point | 0°C |
Boiling Point | 100°C |
CAS Min % | 1%, 99% |
CAS Max % | 99%, 1% |
Concentration | 1% |
Dye Content | 1% |
Assay Percent Range | 1% |
Extinction Coefficient | 171004 M-1cm-1 |
Linear Formula | C15H17N4 |
UN Number | Not regulated. |
Show More |
Safety and Handling
Hazard Category | None. |
ShelfLife | 3 years |
DOTInformation | Not regulated. |
EINECSNumber | 209-035-8 |
RTECSNumber | SG1400000 |
Recommended Storage | Store in a cool, well-ventilated area away from incompatible substances. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction