Learn More
Neurturin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P102BT1MG
Description
AA Sequence of recombinant protein: ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Neurturin. Anti-Human Neurturin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Neurturin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Neurturin (500-P102) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Neurturin. Western Blot: To detect hNeurturin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNeurturin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P102BT-1MG will be provided as 2 x 500 μg
Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.Specifications
Neurturin | |
Polyclonal | |
Biotin | |
NRTN | |
neurturin; NRTN; NTN; prepro-neurturin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4902 | |
-20°C |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
Q99748 | |
NRTN | |
E.coli-derived, 23.6 kDa Recombinant Human Neurturin | |
2 x 500 μg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.