Learn More
Eurofins Discoverx Neuromedin U-25, 500 μg. (92-1174)
Supplier:  Eurofins Discoverx 921174
Encompass_Preferred
Neuromedin U-25 is a 25 amino acid neuropeptide found in the human brain that has functions in smooth muscle contraction, blood pressure regulation, bone growth, pain perception, appetite, and hormone release. Isoforms of different lengths are found in different species. Found to be overexpressed in pancreatic cancers. Activates the neuromedin receptors nMU1R and nMU2R. Provided as 500μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9702
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.