Learn More
STEMCELL Technologies NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat), 1 Kit
Supplier: STEMCELL Technologies 05715
NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat) is recommended for the enzymatic digestion and dissociation of adult mouse and rat central nervous system (CNS) tissue. NeuroCult™ Enzymatic Dissociation Kit has been optimized so that the entire procedure is reproducible, fast, and yields high cell numbers and viabilities. The resulting single-cell suspension is ready for immediate use in downstream applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.