Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Nepicastat hydrochloride
Potent and selective dopamine β-hydroxylase (DBH) inhibitor; orally bioavailable
$231.00 - $965.50
Specifications
Chemical Name or Material | 5-(Aminomethyl)-1-[(2S)-5,7-difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]-1,3-dihydro-2H-imidazole-2-thione hydrochloride |
---|---|
CAS | 170151-24-3 |
Target | Hydroxylase Inhibitors |
Purity | >98% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-371-0RD
|
Tocris Bioscience™
5037/10 |
10 mg |
Each for $231.00
|
|
|||||
50-375-0RD
|
Tocris Bioscience™
5037/50 |
50 mg |
Each for $965.50
|
|
|||||
Description
Potent and selective dopamine β-hydroxylase (DBH) inhibitor (IC50 = 9 nM) . Selective for DBH over 12 other enzymes. Decreases noradrenaline and increases dopamine levels in tissues and plasma in hypertensive rats. Prevents progressive left ventricular dysfunction in an in vivo heart failure model. Also potentiates the stimulus effects of cocaine without producing cocaine-like effects in rats. Orally bioavailable.Specifications
5-(Aminomethyl)-1-[(2S)-5,7-difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]-1,3-dihydro-2H-imidazole-2-thione hydrochloride | |
170151-24-3 | |
Hydroxylase Inhibitors | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title