Learn More
Invitrogen™ Neon™ Transfection Tubes
SDP
The Neon™ Transfection Tubes are designed specifically for use with the Neon™ Transfection System device (MPK5000).
Supplier:  Invitrogen™ MPT100
Encompass
Catalog No. MPT100
Explore all special offers
Add to Cart
Description
Description
Each Neon™ Transfection System Kit (MPK1096 and MPK10096) already comes with 20 electroporation tubes. The additional pack of 100 tubes is offered for those labs wishing to use single tubes per reaction depending on their experimental set-up.
Specifications
Specifications
The Neon™ Transfection Tubes comes in a pack of 100 tubes. Refer to the Neon™ Transfection System users manual for protocol instructions. For use at room temperature under normal laboratory conditions.
RNA, DNA
Transfection
Transfection Tube
Yes
2x10^4 - 6 x10^8 cells; Varies by cell type
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Neon™ Transfection System
Neon™
1 Pack
Room Temperature
Neon™ Transfection System
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only. Not for use in diagnostic procedures.