Learn More
Neogen Diagnostics K-Blue Advanced TMB Substrate, 200 mL, Microwell, HRP, Colorimetric
Supplier:  Neogen Diagnostics 319175
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
K-Blue Advanced is a one-bottle stabilized chromogenic substrate for use with horseradish peroxidase (HRP) immunoassays. K-Blue Advanced offers superior sensitivity, long-term stability and is 100% REACH compliant. K-Blue Advanced develops a deep blue color in the presence of peroxidase labelled conjugate and has been optimized for microwell assay applications, including ELISA. K-Blue Advanced is not applicable for use with assays requiring a precipitating substrate. Stable for a minimum of 36-months when stored at 2-8C.
Catalog No. NC1849069