Learn More
Neogen Diagnostics Estradiol ELISA Kit - 96 well
Supplier:  Neogen Diagnostics 402110
Encompass_Preferred
Estradiol ELISA is used for the quantitative analysis of Estradiol levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. Assay range: 0.02 - 2 ng/mL, Assay sample size: 50 µL.
Catalog No. NC1849082
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.