Learn More
Med Vet International FlipLock Safety Needles, 30G x 0.5in, 50/Box
Supplier: Med Vet International 813005
Perhaps the most versatile and cost-effective safety needle available. EasyTouch FlipLock safety needles fit all standard Luer Lock and Slip Fit syringe barrels. Easy to Use. Protective Cover Shields Needle After Use. Multi-lock Safety System. Latex Free
This item is not returnable.
View return policy