Learn More
New England Biolabs, Inc. NEBNext Multiplex Oligos for Illumina (96 Unique Dual Index Primer Pairs Set 2) – 96 reactions
SDP
Supplier:  New England Biolabs, Inc. E6442S
Encompass
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. Unique Dual Index (UDI) primer pairs enable the identification and mitigation of the impacts of index hopping when using a patterened flow cell. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 96 pre-mixed unique pairs of i5 and i7 index primers, packaged in a single-use 96-well plate with a pierceable foil seal. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Catalog No. 50-202-4009
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.