Learn More
New England Biolabs, Inc. NEBuffer™ 4 – 5 ml
Supplier: New England Biolabs, Inc. B7004S
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. Occasionally, an enzyme has specific buffer requirements not met by one of the four standard NEBuffers, in which case the enzyme is supplied with its own unique NEBuffer. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1 , NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available. Visit www.neb.com for details.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.