Learn More
New England Biolabs, Inc. NEBNext UltraExpress™ FS DNA Library Prep Kit - 24 rxns
Supplier: New England Biolabs, Inc. E3340S
The NEBNext UltraExpress FS DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow, including a fragmentation step. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range of intact DNA, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.