Learn More
New England Biolabs, Inc. NEBNext UltraExpress DNA 96
SDP
Supplier:  New England Biolabs, Inc. E3325L
Encompass
The NEBNext UltraExpress DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.
Catalog No. 50-252-7903
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.