Learn More
New England Biolabs, Inc. NEBNext Ultra™ II End Repair/dA-Tailing Module – 24 reactions
SDP
Supplier:  New England Biolabs, Inc. E7546S
Encompass
This module is part of the Ultra II workflow, and is optimized for use with the NEBNext Ultra II Ligation Module , for Illumina-compatible library construction. This module is also compatible with some Oxford Nanopore MinIONworkflows.
Catalog No. 50-104-7503
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.