Learn More
New England Biolabs, Inc. NEBNext Companion Module for Oxford Nanopore TechnologiesLigation Sequencing
SDP
Supplier:  New England Biolabs, Inc. E7180L
Encompass
The NEBNext DNA repair end repair and ligation reagents recommended in Ligation library preparation are now available in the same product at volumes designed for use in several protocols. The appropriate device-specific protocol from Oxford Nanopore Technologies should be followed for use of these reagents.
Catalog No. 50-225-3072
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.