Learn More
New England Biolabs, Inc. NEBNext Immune Sequencing Kit (Human) - 24 rxn
SDP
Supplier:  New England Biolabs, Inc. E6320S
Encompass
The NEBNext Immune Sequencing Kit (Human) enables exhaustive profiling of somatic mutations in the full-length immune gene repertoires of B cells and T cells, via the expression of complete antibody chains. This includes modular primer sets, providing information for complete V, D, and J segments and full isotype information analysis (IgM, IgD, IgG, IgA and IgE), and TCR(alpha symbol) and TCR(beta symbol) chain characterization. A unique, UMI-based mRNA barcoding process allows PCR copies derived from an individual molecule to be converted to a consensus sequence. This improves sequence accuracy and eliminates PCR bias, allowing for quantitative digital molecule counting. Immune repertoire sequencing is frequently used to analyze immune responses, both current and distant. Areas of particular interest include characterization of autoimmune diseases, oncology, and the discovery of neutralizing antibodies against infectious disease.
Catalog No. 50-210-8560
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.