Learn More
New England Biolabs, Inc. NEBNext Enzymatic 5hmC-seq Conversion Module 96 rxns
SDP
Supplier:  New England Biolabs, Inc. E3365L
Encompass
The NEBNext Enzymatic 5hmC-seq Conversion Module contains the reagents required to enzymatically modify and enable specific detection of 5-hydroxymethylcytosine (5hmC). The user-friendly workflow enables 5hmC identification from 0.1 – 200 ng of input DNA. 
Catalog No. 50-255-1936
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.