Learn More
New England Biolabs, Inc. NEBNext Enzymatic 5hmC-seq Kit 24 rxns
Supplier: New England Biolabs, Inc. E3350S
The NEBNext Enzymatic 5hmC-seq Kit specifically detects 5hmC sites. The kit includes NEBNext Ultra II library prep reagents, and 5hmC is detected using a two-step enzymatic conversion workflow that minimizes damage to DNA and allows discrimination of 5hmC from both cytosine and 5mC after Illumina sequencing. E5hmC-seq data can also be subtracted from EM-seq data, allowing determination of the precise location of individual 5mC and 5hmC sites. The kit includes the E5hmC-seq adaptor and index primers are supplied separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.