Learn More
New England Biolabs, Inc. NEBNext Adaptor Dilution Buffer – 9.6 ml
Supplier: New England Biolabs, Inc. B1430S
The NEBNext Adaptor Dilution Buffer is optimized for dilution of the NEBNext Adaptor to the appropriate concentration range for efficient ligation to end-repaired, dA-tailed DNA.