Learn More
New England Biolabs, Inc. NEBExpress GamS Nuclease Inhibitor – 75 µg
SDP
Supplier:  New England Biolabs, Inc. P0774S
Encompass
NEBExpress GamS Nuclease Inhibitor is a recombinant protein that inhibits Exonuclease V (RecBCD) activity and stabilizes linear DNA templates in E. coli based in vitro protein synthesis reactions.
Catalog No. 50-202-4051
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.