Learn More
New England Biolabs, Inc. NEB Golden Gate Assembly Kit (BsmBI-v2) – 100 reactions
SDP
Supplier:  New England Biolabs, Inc. E1602L
Encompass
The NEB Golden Gate Assembly Kit (BsmBI-v2) contains an optimized mix of BsmBI-v2 and T4 DNA Ligase. BsmBI-v2 has been engineered by NEB and outperforms BsmBI in Golden Gate Assemblies. Together these enzymes can direct the assembly of multiple inserts/modules and also single insert/library generation cloning with single insert(s) using the Golden Gate approach. The pGGAselect destination plasmid provides a backbone for your assembly. It has flanking recognition sites in the correct orientation for BsmBI-directed assemblies, and also BsaI- and BbsI-directed assemblies, enabling the destination plasmid to conveniently be used with the most commonly used Type IIS restriction enzymes used for Golden Gate Assembly. Please note that while general descriptions regarding Golden Gate Assembly use the BsmBI nomenclature, this kit and protocols feature the specific engineered form optimized for Golden Gate Assembly, BsmBI-v2.
Catalog No. 50-202-4002
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.