Learn More
New England Biolabs, Inc. NEB 10-beta Competent E. coli (High Efficiency) – 6 x 0.2 ml/tube
Supplier: New England Biolabs, Inc. C3019I
NEB 10-beta Competent E. coli is a derivative of the popular DH10B. It is T1 phage resistant and endonuclease I (endA1) deficient for high- quality plasmid preparations.