Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBMPR, Tocris Bioscience™

Catalog No. 292450
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Equilibrative nucleoside transporter 1 (ENT1) inhibitor

Equilibrative nucleoside transporter 1 (ENT1) inhibitor (Ki values are 0.4 and 2800 nM for hENT1 and hENT2 respectively).

Chemical Identifiers

CAS 38048-32-7
Molecular Formula C17H17N5O6S
Molecular Weight (g/mol) 419.412
InChI Key DYCJFJRCWPVDHY-LSCFUAHRSA-N
Synonym nbmpr, 4-nitrobenzylthioinosine, s-4-nitrobenzyl-6-thioinosine, nbti, 6-p-nitrobenzylthio inosine, unii-gv1l2dzm2z, gv1l2dzm2z, 6-4-nitrobenzyl thio-9-beta-d-ribofuranosylpurine, s6-nitrobenzyl mercaptopurine riboside, 6-4-nitrophenyl methyl thio-9-beta-d-ribofuranosyl-9h-purine
PubChem CID 65407
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[(4-nitrophenyl)methylsulfanyl]purin-9-yl]oxolane-3,4-diol
SMILES C1=CC(=CC=C1CSC2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)O)O)[N+](=O)[O-]

Specifications

Quantity 50 mg
Formula Weight 419.41
Percent Purity >99%
Chemical Name or Material NBMPR
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.