Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene-1,4-dicarbonitrile, 98+%, Thermo Scientific Chemicals

Catalog No. AAL2024822
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3029-30-9
Molecular Formula C12H6N2
Molecular Weight (g/mol) 178.194
MDL Number MFCD03093626
InChI Key BENSWQOUPJQWMU-UHFFFAOYSA-N
Synonym 1,4-dicyanonaphthalene, 1,4-naphthalenedicarbonitrile, 1,4-dicyanonahthalene, acmc-209heb, ghl.pd_mitscher_leg0.246
PubChem CID 76414
IUPAC Name naphthalene-1,4-dicarbonitrile
SMILES C1=CC=C2C(=CC=C(C2=C1)C#N)C#N

Specifications

Melting Point 208°C to 212°C
Odor Odorless
Quantity 100 g
Beilstein 1870226
Formula Weight 178.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-1,4-dicarbonitrile
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P280f
Wear protective clothing.
EINECSNumber 221-197-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.