Learn More
Enzo Life Sciences NAO (ultra pure) (25mg). CAS: 75168-11-5
Supplier: Enzo Life Sciences ENZ52306
10-N-Nonyl acridine orange (NAO) is an acridine orange derivative and is generally used as a fluorescent marker of the inner mitochondrial membrane in whole cells. It is believed to bind to negatively charged phospholipids. NAO accumulation in the cell seems to be related to specific interactions with mitochondrial membrane proteins and/or lipids, such as cardiolipin, and is largely independent of mitochondrial membrane potential. With respect to apoptosis, the presence of mitochondrial membrane potential can be probed with Rhodamine123 while the structure and integrity of mitochondria can be assessed using 10-N-nonyl-acridine orange. Wavelength Maxima: Excitation 495nm, Emission 519nm Alternative name: Nonyl Acridine Orange, 3,6-bis(Dimethylamino)-10-nonyl-acridinium bromide. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.