Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck Nanotrap Microbiome B Particles
Nanotrap Microbiome B Particles enrich for bacteria including Escherichia coli, Salmonella enterica, Campylobacter jejuni, Listeria monocytogenes, Clostridioides difficile and crASSphage.
$916.00 - $2435.00
Specifications
Isolation Technology | Magnetic Particles |
---|---|
Sample Type | Wastewater |
Sequencing Type | RT-PCR and Digital PCR Sequencing |
Product Type | Microbiome B Particles |
Form | Liquid |
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Price | Quantity & Availability | |||||
23-285-976
|
Streck
250075 |
10 mL |
Each for $916.00
|
|
|||||
23-285-977
|
Streck
250076 |
30 mL |
Each for $2,435.00
|
|
|||||
Description
- Utilize affinity capture and enrichment to attract and bind low abundance viruses.
- Can be used with clinical swab samples, saliva, urine, wastewater, food wash and blackwater.
- Compatible with PCR, sequencing or immunoassay analysis methods.
- Designed to capture and concentrate a variety of microbes.
- Increase sensitivity of existing SARS-CoV-2 assay by capturing and concentrating whole virus.
- Nanotrap Enhancement Reagents improve microbe detection in wastewater samples by at least 1-2 Ct values.
Specifications
Magnetic Particles | |
RT-PCR and Digital PCR Sequencing | |
Liquid |
Wastewater | |
Microbiome B Particles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title