Learn More
Enzo Life Sciences Nadifloxacin (1g). CAS: 124858-35-1
SDP
Supplier:  Enzo Life Sciences ALX380294G001
Encompass_Preferred
Fluoroquinolone with broad spectrum activity against Gram-negative and Gram-positive bacteria. Inhibits the generation of O2- and OH radicals by neutrophils. Inhibits the production of inflammatory cytokines. Alternative name: 9-Fluoro-6,7-dihydro-8-(4-hydroxypiperidino)-5-methyl-1-oxo-1H,5H-benzo[ij]quinolizine-2-carboxylic acid, Jinofloxacin, OPC-7251. Purity: ≥98% (Assay). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9020
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.