Learn More
N-Ras Recombinant Rabbit Monoclonal Antibody (10H6L11), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES 703435
Description
This antibody is predicted to react with Pig, Rat, Rabbit. Recombit rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombit rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noo syndrome 6.Specifications
N-Ras | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01111 | |
NRAS | |
Peptides corresponding to human N-Ras [1) aa83-aa98 2) aa166-aa180] | |
100μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
10H6L11 | |
Unconjugated | |
NRAS | |
ALPS4; AV095280; CMNS; GTPase NRas; HRAS1; NCMS; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; Nras; N-ras; N-ras protein part 4; NRAS1; NS6; RAS; RASN; transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog | |
Rabbit | |
Protein A | |
RUO | |
4893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.