Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl methacrylate, 95%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog No. AAL0949314
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2210-28-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00048690
InChI Key NHARPDSAXCBDDR-UHFFFAOYSA-N
Synonym propyl methacrylate, n-propyl methacrylate, methacrylic acid, propyl ester, propyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, propyl ester, unii-95q6j7a6n3, ccris 4864, methacrylic acid propyl, propyl 2-methylacrylate, propyl 2-methylacrylate #
PubChem CID 16638
IUPAC Name propyl 2-methylprop-2-enoate
SMILES CCCOC(=O)C(C)=C

Specifications

Density 0.902
Boiling Point 140°C to 141°C
Flash Point 30°C (86°F)
Odor Pungent
Refractive Index 1.419
Quantity 25 g
UN Number UN3272
Beilstein 1744849
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material n-Propyl methacrylate, stabilized with 200ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 218-639-0
RTECSNumber OZ5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.