Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Phenylmaleimide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Melting Point | 87°C to 91°C |
Boiling Point | 162°C to 163°C (12 mmHg) |
Quantity | 500 g |
UN Number | UN2811 |
Beilstein | 125098 |
Merck Index | 14,7299 |
Solubility Information | Soluble in water (slightly ),methanol,ethanol,and benzene. |
Formula Weight | 173.17 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Phenylmaleimide |
Safety and Handling
Hazard Category | H301-H315-H319 |
Hazard Statement | GHS H Statement H301-H319 Toxic if swallowed. Causes serious eye irritation. |
Precautionary Statement | P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 213-382-0 |
RTECSNumber | ON5950000 |
TSCA | Yes |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction